Assay Details
| Gene Symbol: CDKN1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100346.1 | NP_001093816.1 | 1 - 2 | 991 | 77 | |
| GenBank mRNA | BC148067.1 | - | 1 - 2 | 991 | 77 | |
More Information
Set Membership: |
|

CDKN1B
Genomic Map