Assay Details
| Gene Symbol: EBI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100365.2 | NP_001093835.1 | 1 - 2 | 99 | 59 | |
| GenBank mRNA | BC149502.1 | - | 1 - 2 | 100 | 59 | |
More Information
Set Membership: |
|

EBI3
Genomic Map