Assay Details
Gene Symbol: NAAA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100369.2 | NP_001093839.1 | 2 - 3 | 433 | 61 | |
GenBank mRNA | BC149381.1 | - | 2 - 3 | 435 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
