Assay Details
| Gene Symbol: SMPDL3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100379.1 | NP_001093849.1 | 5 - 6 | 874 | 63 | |
| GenBank mRNA | BC149924.1 | - | 5 - 6 | 874 | 63 | |
More Information
Set Membership: |
|

SMPDL3B
Genomic Map