Assay Details
Gene Symbol: SLC31A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100381.1 | NP_001093851.1 | 2 - 3 | 239 | 102 | |
GenBank mRNA | BC150134.1 | - | 2 - 3 | 239 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
