Assay Details
| Gene Symbol: IARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101069.2 | NP_001094539.1 | 132 | |||
| GenBank mRNA | BC151484.1 | - | 132 | |||
| BC154383.1 | - | 132 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IARS1| Gene Symbol: IARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101069.2 | NP_001094539.1 | 132 | |||
| GenBank mRNA | BC151484.1 | - | 132 | |||
| BC154383.1 | - | 132 | ||||