Assay Details
| Gene Symbol: IARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101069.2 | NP_001094539.1 | 3 - 4 | 346 | 65 | |
| GenBank mRNA | BC151484.1 | - | 3 - 4 | 347 | 65 | |
More Information
Set Membership: |
|

IARS1
Genomic Map