Assay Details
| Gene Symbol: L2HGDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101090.1 | NP_001094560.1 | 5 - 6 | 771 | 78 | |
| GenBank mRNA | BC151577.1 | - | 5 - 6 | 771 | 78 | |
More Information
Set Membership: |
|

L2HGDH
Genomic Map