Assay Details
| Gene Symbol: KCTD20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101093.1 | NP_001094563.1 | 3 - 4 | 556 | 68 | |
| GenBank mRNA | BC151360.1 | - | 3 - 4 | 556 | 68 | |
More Information
Set Membership: |
|

KCTD20
Genomic Map