Assay Details
| Gene Symbol: PACSIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101101.1 | NP_001094571.1 | 6 - 7 | 971 | 64 | |
| GenBank mRNA | BC151572.1 | - | 6 - 7 | 971 | 64 | |
| BC153241.1 | - | 5 - 6 | 840 | 64 | ||
More Information
Set Membership: |
|

PACSIN1
Genomic Map