Assay Details
Gene Symbol: TRIB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101105.2 | NP_001094575.1 | 2 - 3 | 1161 | 107 | |
GenBank mRNA | BC147945.1 | - | 2 - 3 | 1163 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
