Assay Details
| Gene Symbol: TADA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101106.1 | NP_001094576.1 | 3 - 4 | 336 | 79 | |
| GenBank mRNA | BC150064.1 | - | 3 - 4 | 336 | 79 | |
More Information
Set Membership: |
|

TADA1
Genomic Map