Assay Details
| Gene Symbol: NEDD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101847.4 | NP_001095317.4 | 1 - 2 | 962 | 94 | |
| GenBank mRNA | BC149241.1 | - | 2 - 3 | 516 | 94 | |
More Information
Set Membership: |
|

NEDD9
Genomic Map