Assay Details
Gene Symbol: KRT20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101851.2 | NP_001095321.1 | 6 - 7 | 1189 | 71 | |
GenBank mRNA | BC149955.1 | - | 6 - 7 | 1191 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
