Assay Details
| Gene Symbol: RBM48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101854.2 | NP_001095324.2 | 3 - 4 | 467 | 141 | |
| GenBank mRNA | BC149271.1 | - | 2 - 3 | 279 | 141 | |
| DV784940.1 | - | 490 | 141 | |||
More Information
Set Membership: |
|

RBM48
Genomic Map