Assay Details
Gene Symbol: SUSD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101904.1 | NP_001095374.1 | 5 - 6 | 836 | 54 | |
GenBank mRNA | BC148046.1 | - | 5 - 6 | 836 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
