Assay Details
| Gene Symbol: MFSD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101905.2 | NP_001095375.2 | 3 - 4 | 1198 | 114 | |
| GenBank mRNA | BC148909.1 | - | 3 - 4 | 1199 | 114 | |
More Information
Set Membership: |
|

MFSD3
Genomic Map