Assay Details
| Gene Symbol: ACSM2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101952.2 | NP_001095422.1 | 7 - 8 | 1085 | 75 | |
| GenBank mRNA | BC151513.1 | - | 7 - 8 | 1086 | 75 | |
More Information
Set Membership: |
|

ACSM2B
Genomic Map