Assay Details
| Gene Symbol: HERC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101995.1 | NP_001095465.1 | 16 - 17 | 2309 | 78 | |
| GenBank mRNA | BC149081.1 | - | 16 - 17 | 2309 | 78 | |
More Information
Set Membership: |
|

HERC5
Genomic Map