Assay Details
| Gene Symbol: SPAG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102005.2 | NP_001095475.1 | 1 - 2 | 82 | 69 | |
| GenBank mRNA | BC149514.1 | - | 1 - 2 | 83 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC149514.1, NM_001102005.2 transcripts.
Set Membership: |
|

SPAG8
Genomic Map