Assay Details
| Gene Symbol: TCAIM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102016.1 | NP_001095486.1 | 5 - 6 | 875 | 64 | |
| GenBank mRNA | BC149840.1 | - | 5 - 6 | 875 | 64 | |
More Information
Set Membership: |
|

TCAIM
Genomic Map