Assay Details
| Gene Symbol: LOC515697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102030.2 | NP_001095500.1 | 2 - 3 | 101 | 69 | |
| GenBank mRNA | BC149652.1 | - | 2 - 3 | 102 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC149652.1, NM_001102030.2 transcripts.
Set Membership: |
|

LOC515697
Genomic Map