Assay Details
Gene Symbol: PI4K2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102064.1 | NP_001095534.1 | 3 - 4 | 773 | 85 | |
GenBank mRNA | BC147892.1 | - | 3 - 4 | 773 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
