Assay Details
| Gene Symbol: KRT78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102077.1 | NP_001095547.1 | 2 - 3 | 614 | 58 | |
| GenBank mRNA | BC149049.1 | - | 2 - 3 | 614 | 58 | |
More Information
Set Membership: |
|

KRT78
Genomic Map