Assay Details
Gene Symbol: SEPTIN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102087.2 | NP_001095557.1 | 1 - 2 | 35 | 150 | |
GenBank mRNA | BC149885.1 | - | 1 - 2 | 37 | 150 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC149885.1, NM_001102087.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
