Assay Details
Gene Symbol: SEPTIN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102087.2 | NP_001095557.1 | 3 - 4 | 232 | 144 | |
GenBank mRNA | BC149885.1 | - | 3 - 4 | 234 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
