Assay Details
Gene Symbol: SEPTIN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102087.2 | NP_001095557.1 | 7 - 8 | 873 | 76 | |
GenBank mRNA | BC149885.1 | - | 7 - 8 | 875 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
