Assay Details
Gene Symbol: SEPTIN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102087.2 | NP_001095557.1 | 8 - 9 | 1038 | 89 | |
GenBank mRNA | BC149885.1 | - | 8 - 9 | 1040 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
