Assay Details
| Gene Symbol: AKR1B10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102088.2 | NP_001095558.1 | 2 - 3 | 324 | 71 | |
| GenBank mRNA | BC151501.1 | - | 2 - 3 | 325 | 71 | |
More Information
Set Membership: |
|

AKR1B10
Genomic Map