Assay Details
Gene Symbol: PCDHA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102506.2 | NP_001095976.1 | 1 - 2 | 2517 | 107 | |
GenBank mRNA | BC151664.1 | - | 1 - 2 | 2518 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
