Assay Details
| Gene Symbol: CBX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103094.1 | NP_001096564.1 | 2 - 3 | 121 | 77 | |
| GenBank mRNA | AY962102.1 | - | 1 - 2 | 51 | 77 | |
| BC133330.1 | - | 2 - 3 | 121 | 77 | ||
More Information
Set Membership: |
|

CBX6
Genomic Map