Assay Details
| Gene Symbol: CBX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103094.1 | NP_001096564.1 | 4 - 5 | 259 | 77 | |
| GenBank mRNA | AY962102.1 | - | 3 - 4 | 189 | 77 | |
| BC133330.1 | - | 4 - 5 | 259 | 77 | ||
More Information
Set Membership: |
|

CBX6
Genomic Map