Assay Details
| Gene Symbol: FITM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103095.1 | NP_001096565.1 | 1 - 2 | 223 | 70 | |
| GenBank mRNA | BC134674.1 | - | 1 - 2 | 223 | 70 | |
More Information
Set Membership: |
|

FITM2
Genomic Map