Assay Details
Gene Symbol: ZNF404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104965.1 | NP_001098435.1 | 2 - 3 | 263 | 147 | |
GenBank mRNA | BC149662.1 | - | 2 - 3 | 263 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
