Assay Details
| Gene Symbol: PRODH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104967.2 | NP_001098437.1 | 9 - 10 | 1289 | 64 | |
| GenBank mRNA | BC149682.1 | - | 9 - 10 | 1290 | 64 | |
More Information
Set Membership: |
|

PRODH2
Genomic Map