Assay Details
Gene Symbol: CDCA7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104977.1 | NP_001098447.1 | 7 - 8 | 1104 | 72 | |
GenBank mRNA | BT030712.1 | - | 7 - 8 | 1104 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
