Assay Details
| Gene Symbol: TDP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105341.1 | NP_001098811.1 | 6 - 7 | 855 | 87 | |
| GenBank mRNA | BC134603.1 | - | 6 - 7 | 855 | 87 | |
More Information
Set Membership: |
|

TDP2
Genomic Map