Assay Details
| Gene Symbol: CD8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105344.2 | NP_001098814.1 | 5 - 6 | 671 | 64 | |
| GenBank mRNA | BC134439.1 | - | 5 - 6 | 674 | 64 | |
More Information
Set Membership: |
|

CD8B
Genomic Map