Assay Details
| Gene Symbol: COMTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105368.2 | NP_001098838.1 | 3 - 4 | 446 | 74 | |
| GenBank mRNA | BC134766.1 | - | 3 - 4 | 447 | 74 | |
More Information
Set Membership: |
|

COMTD1
Genomic Map