Assay Details
| Gene Symbol: DTX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105393.1 | NP_001098863.1 | 3 - 4 | 886 | 73 | |
| GenBank mRNA | BC153272.1 | - | 3 - 4 | 886 | 73 | |
More Information
Set Membership: |
|

DTX3
Genomic Map