Assay Details
| Gene Symbol: RSPO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105621.1 | NP_001099091.1 | 3 - 4 | 833 | 55 | |
| GenBank mRNA | BC149052.1 | - | 3 - 4 | 833 | 55 | |
| BT030732.1 | - | 1 - 2 | 158 | 55 | ||
More Information
Set Membership: |
|

RSPO1
Genomic Map