Assay Details
| Gene Symbol: CYP2C19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109792.2 | NP_001103262.1 | 3 - 4 | 500 | 74 | |
| GenBank mRNA | BC153845.1 | - | 3 - 4 | 502 | 74 | |
More Information
Set Membership: |
|

CYP2C19
Genomic Map