Assay Details
| Gene Symbol: MCRIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109794.2 | NP_001103264.1 | 2 - 3 | 116 | 150 | |
| GenBank mRNA | BC149379.1 | - | 2 - 3 | 117 | 150 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC149379.1, NM_001109794.2 transcripts.
Set Membership: |
|

MCRIP1
Genomic Map