Assay Details
Gene Symbol: KIF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110788.1 | NP_001104258.1 | 7 - 8 | 2110 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
