Assay Details
| Gene Symbol: ATP5MF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113719.2 | NP_001107191.1 | 252 | 85 | ||
| GenBank mRNA | BC108208.1 | - | 253 | 85 | ||
| S70447.1 | - | 3 - 4 | 252 | 85 | ||
More Information
Set Membership: |
|

ATP5MF
Genomic Map