Assay Details
| Gene Symbol: PLA2G4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114657.1 | NP_001108129.1 | 5 - 6 | 473 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PLA2G4B| Gene Symbol: PLA2G4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114657.1 | NP_001108129.1 | 5 - 6 | 473 | 77 | |
Set Membership: |
|