Assay Details
| Gene Symbol: DUSP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034725.2 | NP_001029897.2 | 3 - 4 | 1317 | 63 | |
| GenBank mRNA | BC122694.1 | - | 3 - 4 | 1317 | 63 | |
| BT021576.1 | - | 3 - 4 | 1324 | 63 | ||
More Information
Set Membership: |
|

DUSP10
Genomic Map