Assay Details
Gene Symbol: KCTD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035421.2 | NP_001030498.2 | 1 | 49 | 59 | |
GenBank mRNA | DV877305.1 | - | 1 | 49 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001035421.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
