Assay Details
Gene Symbol: WTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046341.3 | NP_001039806.1 | 2 - 3 | 218 | 75 | |
NM_001113254.2 | NP_001106725.1 | 2 - 3 | 218 | 75 | ||
GenBank mRNA | BC112754.1 | - | 2 - 3 | 204 | 75 | |
DV801115.1 | - | 2 - 3 | 209 | 75 | ||
DY142750.1 | - | 1 | 149 | 75 | ||
EE372654.1 | - | 2 - 3 | 61 | 75 | ||
EH196049.1 | - | 2 - 3 | 219 | 75 |
Gene Symbol: LOC104975673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC149777.1 | - | 2 | 321 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC112754.1, NM_001046341.3, NM_001113254.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
