Assay Details
Gene Symbol: WTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046341.3 | NP_001039806.1 | 3 - 4 | 268 | 75 | |
NM_001113254.2 | NP_001106725.1 | 3 - 4 | 268 | 75 | ||
GenBank mRNA | BC112754.1 | - | 3 - 4 | 254 | 75 | |
DV801115.1 | - | 3 - 4 | 259 | 75 | ||
DY142750.1 | - | 1 | 199 | 75 | ||
EE372654.1 | - | 3 - 4 | 111 | 75 |
Gene Symbol: LOC104975673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC149777.1 | - | 2 | 371 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
